Learn More
Invitrogen™ KCTD14 Polyclonal Antibody

Description
Immunogen sequence: RPILDYLRTG QVPTQHIPEV YREAQFYEIK PLVKLLEDMP QIFGEQVSRK QFLLQVPGYS ENLELMVRLA RAEAITARKS SVLV Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 70%.
Specifications
Specifications
| Antigen | KCTD14 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | KCTD14 |
| Gene Accession No. | Q9BQ13 |
| Gene Alias | AI449310; BTB/POZ domain-containing protein KCTD14; D7Ertd760e; KCTD14; potassium channel tetramerisation domain containing 14; potassium channel tetramerization domain containing 14 |
| Gene Symbols | KCTD14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.