Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD15 Antibody (2B11), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260232APC
This item is not returnable.
View return policy
Description
KCTD15 Monoclonal antibody specifically detects KCTD15 in Human samples. It is validated for Flow CytometrySpecifications
KCTD15 | |
Monoclonal | |
APC | |
BTB/POZ domain-containing protein KCTD15, MGC25497, MGC2628, potassium channel tetramerisation domain containing 15 | |
Recombinant human KCTD15(1-159aa) purified from E. coli. | |
0.1 mL | |
metabolism | |
79047 | |
Store at 4°C in the dark. | |
IgG2b κ |
Flow Cytometry | |
2B11 | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction