Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD15 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177007MFV500
Description
KCTD15 Polyclonal antibody specifically detects KCTD15 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
KCTD15 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
BTB/POZ domain-containing protein KCTD15, MGC25497, MGC2628, potassium channel tetramerisation domain containing 15 | |
Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human KCTD15. The immunogen is located within the last 50 amino acids of KCTD15. Amino Acid Squence: REEERRPQPTPTAVR | |
0.1 mL | |
metabolism | |
79047 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction