Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCTD16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560766
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560766 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560766 Supplier Invitrogen™ Supplier No. PA560766
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDLVKLLTPD EIKQSPDEFC HSDFEDASQG SDTRICPPSS LLPADRKWGF ITVGYRGSCT LGREGQADAK F Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 33%.

The BTB (Broad-Complex, Tramtrack and Bric a brac) domain, also known as the POZ (Poxvirus and Zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. KCTD16 (potassium channel tetramerisation domain containing 16), also known as BTB/POZ domain-containing protein KCTD16, is a 428 amino acid protein that contains one BTB (POZ) domain. An auxiliary subunit of GABAB R1 and GABAB R2, KCTD16 increases agonist potency and alters the G-protein signaling of the receptors by accelerating onset and promoting desensitization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD16
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCTD16
Gene Accession No. Q68DU8
Gene Alias 4930434H12Rik; BTB/POZ domain-containing protein KCTD16; Gm1267; KCTD16; KIAA1317; potassium channel tetramerisation domain containing 16; potassium channel tetramerization domain containing 16; potassium channel tetramerization domain-containing protein 16
Gene Symbols KCTD16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCTD16. Recombinant protein control fragment (Product #RP-96972).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57528
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.