Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCTD17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568686
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568686 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568686 Supplier Invitrogen™ Supplier No. PA568686
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Dog: 79%; Guinea Pig: 92%; Human: 100%; Mouse: 86%; Rat: 86%.

Is a positive regulator of ciliogenesis, playing a crucial role in the initial steps of axoneme extension. It acts as a substrate-adapter for CUL3-RING ubiquitin ligase complexes which mediate the ubiquitination and subsequent proteasomal degradation of TCHP, a protein involved in ciliogenesis down-regulation (PubMed:25270598). May be involved in endoplasmic reticulum calcium ion homeostasis (PubMed:25983243).
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD17
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KCTD17
Gene Accession No. Q8N5Z5
Gene Alias 2900008M13Rik; AA414907; AW742389; BTB/POZ domain-containing protein KCTD17; KCTD17; LOW QUALITY PROTEIN: BTB/POZ domain-containing protein KCTD17; N28155; potassium channel tetramerisation domain containing 17; potassium channel tetramerization domain containing 17; RGD1311154
Gene Symbols KCTD17
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human KCTD17.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79734
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.