Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCTD18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568692
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568692 Supplier Invitrogen™ Supplier No. PA568692
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 86%; Rat: 93%.

KCTD18 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include protein homooligomerization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD18
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KCTD18
Gene Accession No. Q6PI47
Gene Alias 4932411A20Rik; 6530404F10Rik; BTB/POZ domain-containing protein KCTD18; KCTD18; potassium channel tetramerisation domain containing 18; potassium channel tetramerization domain containing 18; RGD1564820; similar to potassium channel tetramerisation domain containing 18
Gene Symbols KCTD18
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human KCTD18.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130535
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.