Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCTD19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532114
Description
Recommended positive controls: U87-MG, SK-N-SH. Predicted reactivity: Mouse (96%), Rat (97%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The KCTD19 (Potassium Channel Tetramerization Domain Containing 19) gene encodes a protein that plays a crucial role in meiotic chromosome organization and male fertility. KCTD19 is located on chromosome 5 in humans. The protein is known to interact with zinc finger protein 541 (ZFP541) and histone deacetylase 1 (HDAC1), highlighting its role in chromatin organization and transcription regulation during meiosis. Mutations or loss-of-function variants in KCTD19 have been implicated in non-obstructive azoospermia (NOA) and male infertility, as evidenced by studies in both human and mouse models. The gene's function in regulating the individualization of meiotic chromosomes is critical, and disruptions can lead to severe meiotic defects and spermatogenic failure.
Specifications
| KCTD19 | |
| Polyclonal | |
| Unconjugated | |
| KCTD19 | |
| 4922504H04Rik; BTB/POZ domain-containing protein KCTD19; KCTD19; potassium channel tetramerisation domain containing 19; potassium channel tetramerization domain containing 19; RGD1561326; testicular tissue protein Li 101 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 146212, 279499 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q17RG1, Q562E2 | |
| KCTD19 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human KCTD19. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction