Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD20 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | KCTD20 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Description
KCTD20 Polyclonal specifically detects KCTD20 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
KCTD20 | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
222658 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LAEDIKGSCFQSGNKRNHEPFIAPERFGNSSVGFGSNSHSQAPEKVTLLVDGTRFVVNPQIFTAHPDTML | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Polyclonal | |
Rabbit | |
Human | |
BTB/POZ domain-containing protein KCTD20, potassium channel tetramerisation domain containing 20 | |
KCTD20 | |
IgG | |
Affinity Purified | |
Specificity of human KCTD20 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title