Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD3 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP244277AF594
Description
KCTD3 Polyclonal antibody specifically detects KCTD3 in Human samples. It is validated for Western BlotSpecifications
| KCTD3 | |
| Polyclonal | |
| Western Blot | |
| BTB/POZ domain-containing protein KCTD3, potassium channel tetramerisation domain containing 3, renal carcinoma antigen NY-REN-45 | |
| The epitope recognized by this antibody maps to a region between residue 675 to 725 of human BTB/POZ domain-containing protein KCTD3 using the numbering given in entry Q9Y597.2 (GeneID 51133). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 51133 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction