Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCTD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559272
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559272 Supplier Invitrogen™ Supplier No. PA559272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNFLRNGELL LPEGFRENQL LAQEAEFFQL KGLAEEVKSR WEKEQLTPRE TTFLEITDNH DRSQGLRIFC NAPDF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

The KCTD4 (Potassium Channel Tetramerization Domain Containing 4) gene encodes a protein involved in various cellular processes, including the regulation of calcium homeostasis and modulation of fibroblast activity. KCTD4 interacts with chloride intracellular channel 1 (CLIC1) to disrupt calcium homeostasis, leading to increased fibronectin secretion, which activates fibroblasts via paracrine signaling. This activation can promote cancer cell invasion through matrix metalloproteinase 24 (MMP24) signaling. The interaction between KCTD4 and CLIC1 presents a potential target for therapeutic strategies, as inhibition of this interaction has been shown to significantly suppress cancer metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCTD4
Gene Accession No. Q8WVF5
Gene Alias bA321C24.3; BTB/POZ domain-containing protein KCTD4; KCTD4; potassium channel tetramerisation domain containing 4; potassium channel tetramerization domain containing 4
Gene Symbols KCTD4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCTD4. Recombinant protein control fragment (Product #RP-98002).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 386618
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.