Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD5 Antibody (OTI3C8), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP271955UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271955UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271955UV Supplier Novus Biologicals Supplier No. NBP271955UV
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KCTD5 Monoclonal specifically detects KCTD5 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen KCTD5
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3C8
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BTB/POZ domain-containing protein KCTD5, FLJ20040, potassium channel tetramerisation domain containing 5
Gene Symbols KCTD5
Host Species Mouse
Immunogen Full length recombinant protein of human KCTD5 (NP_061865) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 54442.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.