Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD5 Antibody (OTI3C8), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271955IR
Description
KCTD5 Monoclonal specifically detects KCTD5 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
KCTD5 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KCTD5 | |
Full length recombinant protein of human KCTD5 (NP_061865) produced in HEK293T cells. | |
0.1 mL | |
54442.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3C8 | |
Western Blot, Immunohistochemistry | |
BTB/POZ domain-containing protein KCTD5, FLJ20040, potassium channel tetramerisation domain containing 5 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction