Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD5 Antibody (OTI3C8), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271955MFV610
Description
KCTD5 Monoclonal antibody specifically detects KCTD5 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow CytometrySpecifications
KCTD5 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
54442 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
OTI3C8 | |
Flow Cytometry | |
BTB/POZ domain-containing protein KCTD5, FLJ20040, potassium channel tetramerisation domain containing 5 | |
Full length human recombinant protein of human KCTD5 (NP_061865) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction