Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCTD5 Monoclonal Antibody (OTI3C8)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525069

Catalog No. PIMA525069


Only null left
Add to Cart

Description

Description

KCTD5 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

KCTD5 is involved in interaction with CUL3 suggests that it may act as a substrate adapter in some E3 ligase complex. Does not affect the function of Kv channel Kv2.1/KCNB1, Kv1.2/KCNA2, Kv4.2/KCND2 and Kv3.4/KCNC4.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KCTD5
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NXV2
KCTD5
Full length human recombinant protein of KCTD5 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2b
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI3C8
Unconjugated
KCTD5
2610030N08Rik; BTB/POZ domain-containing protein KCTD5; KCTD5; mKIAA0176; potassium channel tetramerisation domain containing 5; potassium channel tetramerization domain containing 5
Mouse
Affinity Chromatography
RUO
54442
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.