Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD5 Mouse anti-Human, Clone: OTI3C8, liquid, TrueMAB™
SDP

Catalog No. 501672178
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-2178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-2178 Supplier OriGene Supplier No. TA501035
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KCTD5 is involved in interaction with CUL3 suggests that it may act as a substrate adapter in some E3 ligase complex. Does not affect the function of Kv channel Kv2.1/KCNB1, Kv1.2/KCNA2, Kv4.2/KCND2 and Kv3.4/KCNC4.

Specifications

Antigen KCTD5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KCTD5
Gene Accession No. Q9NXV2
Gene Alias potassium channel tetramerization domain containing 5
Gene Symbols KCTD5
Host Species Mouse
Immunogen Full length human recombit protein of human KCTD5 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54442
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.