Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCTD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557519
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557519 Supplier Invitrogen™ Supplier No. PA557519
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QIEPLIQCLN DPKPLYPMDT FEEVVELSST RKLSKYSNPV AVIITQLTIT TKVHSLLEGI SNYFTKWNKH M Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.

KCTD6 is a domain of potassium channel.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCTD6
Gene Accession No. Q8NC69
Gene Alias 5430433B02Rik; AU044285; BTB/POZ domain-containing protein KCTD6; hypothetical protein LOC436601; KCASH3; KCASH3 protein; KCTD6; kctd6b; potassium channel tetramerisation domain containing 6; potassium channel tetramerisation domain containing 6b; potassium channel tetramerization domain containing 6; potassium channel tetramerization domain containing 6b; Potassium channel tetramerization domain-containing protein 6; zgc:91884
Gene Symbols KCTD6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCTD6. Recombinant protein control fragment (Product #RP-96433).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200845
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.