Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCTD6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541069
Description
Peptide sequence: HLYTTSLTTL TRYPDSMLGA MFGGDFPTTR DPQGNYFINR DGPLFRYVLN Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 93%; Mouse: 93%; Rabbit: 93%; Rat: 93%; Zebrafish: 93%.
KCTD6 is a domain of potassium channel.
Specifications
KCTD6 | |
Polyclonal | |
Unconjugated | |
KCTD6 | |
5430433B02Rik; AU044285; BTB/POZ domain-containing protein KCTD6; hypothetical protein LOC436601; KCASH3; KCASH3 protein; KCTD6; kctd6b; potassium channel tetramerisation domain containing 6; potassium channel tetramerisation domain containing 6b; potassium channel tetramerization domain containing 6; potassium channel tetramerization domain containing 6b; Potassium channel tetramerization domain-containing protein 6; zgc:91884 | |
Rabbit | |
Protein A | |
RUO | |
200845 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NC69 | |
KCTD6 | |
Synthetic peptide directed towards the N-terminal of human KCTD6. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction