Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCTD7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541068
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541068 Supplier Invitrogen™ Supplier No. PA541068
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VVVTGREPDS RRQDGAMSSS DAEDDFLEPA TPTATQAGHA LPLLPQEFPE Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

The KCTD gene family, including KCTD7, encode predicted proteins that contain N-terminal domain that is homologous to the T1 domain in voltage-gated potassium channels (see KCNA1; MIM 176260). KCTD7 displays a primary sequence and hydropathy profile indicating intracytoplasmic localization. EST database analysis showed that KCTD7 is expressed in human and mouse brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KCTD7
Gene Accession No. Q96MP8
Gene Alias 4932409E18; 9430010P06Rik; BTB/POZ domain-containing protein KCTD7; CLN14; EPM3; FLJ32069; KCTD7; potassium channel tetramerisation domain containing 7; potassium channel tetramerization domain containing 7
Gene Symbols KCTD7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human KCTD7 (aa 2-51).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154881
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.