Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCTD8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556781
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556781 Supplier Invitrogen™ Supplier No. PA556781
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEASTPQDNP SSAQQATAHQ PNTLTLDRPS KKAPVQWIPP PDKRRNSELF QTLISKSRET NLSK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 31%.

Auxiliary subunit of GABA-B receptors that determine the pharmacology and kinetics of the receptor response. Increases agonist potency and markedly alter the G-protein signaling of the receptors by accelerating onset and promoting desensitization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD8
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCTD8
Gene Accession No. Q6ZWB6
Gene Alias A730087N02Rik; BTB/POZ domain-containing protein KCTD8; KCTD8; potassium channel tetramerisation domain containing 8; potassium channel tetramerization domain containing 8
Gene Symbols KCTD8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCTD8. Recombinant protein control fragment (Product #RP-95719).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 386617
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.