Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD9 Antibody (OTI2G8), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271968AF350
Description
KCTD9 Monoclonal specifically detects KCTD9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
KCTD9 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
KCTD9 | |
Recombinant protein fragment corresponding to amino acids 1-223 of human KCTD9 (NP_060104) produced in E.coli. | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2G8 | |
Western Blot, Immunohistochemistry | |
BTB/POZ domain-containing protein KCTD9, FLJ20038, potassium channel tetramerisation domain containing 9 | |
Mouse | |
0.1 mL | |
54793 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction