Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD9 Antibody (OTI2G8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245488
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
KCTD9 Monoclonal specifically detects KCTD9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
KCTD9 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
BTB/POZ domain-containing protein KCTD9, FLJ20038, potassium channel tetramerisation domain containing 9 | |
Mouse | |
42.4 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
OTI2G8 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
KCTD9 | |
Human recombinant protein fragment corresponding to amino acids 1-223 of human KCTD9(NP_060104) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
54793 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction