Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCTD9 Recombinant Rabbit Monoclonal Antibody (JE64-67)
GREENER_CHOICE

Catalog No. PIMA544766
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544766 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544766 Supplier Invitrogen™ Supplier No. MA544766
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

KCTD9 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Substrate-specific adapter of a BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex, which mediates the ubiquitination of target proteins, leading to their degradation by the proteasome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE64-67
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Kctd9
Gene Accession No. Q7L273
Gene Alias BTB/POZ domain-containing protein KCTD9; BTBD27; KCTD9; potassium channel tetramerisation domain containing 9; potassium channel tetramerization domain containing 9
Gene Symbols Kctd9
Host Species Rabbit
Immunogen Synthetic peptide within human KCTD9 aa 51-100/389.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54793
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.