Learn More
KD Scientific Centrifan™ PE Personal Evaporator/Condenser Accessory, Rotors
For use with Centrifan™ PE
Specifications
Type Centrifan Rotor
For Use With (Equipment) Centrifan™ PE
View More Specs
Products 7
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
14-831-601
View Documents
KD Scientific
788400
6 x 20 mL Scintillation vial and 1 Polypropylene splitter N/A N/A N/A
14-831-602
View Documents
KD Scientific
788401
8 x 1.6 mL Microcentrifuge tube and 1 Polypropylene splitter N/A N/A N/A
14-831-603
View Documents
KD Scientific
788402
6 x 16 x 100 mm Test tube and 1 Polypropylene splitter N/A N/A N/A
14-831-604
View Documents
KD Scientific
788403
8 x 4 mL Vial and 1 Polypropylene splitter N/A N/A N/A
14-831-605
View Documents
KD Scientific
788404
6 x 30 mL Scintillation vial and 1 Polypropylene splitter N/A N/A N/A
14-831-607
View Documents
KD Scientific
788406
10 x 1.5 mL HPLC vial and 1 Polypropylene splitter N/A N/A N/A
14-831-606
missing translation for 'viewDocuments'
KD Scientific
788405
4 x 40 mL Scintillation vial and 1 Polypropylene splitter N/A N/A N/A
Description
Description
Specifications
Specifications
Centrifan Rotor
Centrifan™ PE
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.