Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KD Scientific Stainless-Steel Syringes

Catalog No. 1493140
Click to view available options
Diameter (Metric) Outside:
12.7 mm
22.3 mm
31.7 mm
38.1 mm
47 mm
Maximum Pressure:
1500 psi
7000 psi
Type:
Luer-Lok Syringe
Swagelok 1/16 in. Syringe
Swagelok 1/4 in. Syringe
Swagelok 1/8 in. Syringe
Capacity (Metric):
2.5 mL
8 mL
20 mL
50 mL
100 mL
200 mL

Durable, electron-beam construction withstands high force, high pressure applications

Eliminates hazards of glass syringe breakage.

  • Compatible with most syringe pumps
  • Fully autoclavable for reuse
  • Adaptable to Luer Lock or Swagelok™ fittings
  • Resistant to most chemicals

Specifications

Autoclavable Autoclavable
Capacity (Metric) 20 mL
Diameter (English) Inside 3/4 in.
Diameter (English) Outside 22/25 in.
Diameter (Metric) Outside 22.3 mm
Diameter (Metric) Inside 19 mm
For Use With Syringe Pump
IP Rating None
Material Stainless Steel (316)
Type Swagelok 1/4 in. Syringe
Maximum Pressure 1500 psi
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.