Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KD Scientific Stainless-Steel Syringes
Click to view available options
Diameter (Metric) Outside:
12.7 mm
22.3 mm
31.7 mm
38.1 mm
47 mm
Maximum Pressure:
1500 psi
7000 psi
Type:
Luer-Lok Syringe
Swagelok 1/16 in. Syringe
Swagelok 1/4 in. Syringe
Swagelok 1/8 in. Syringe
Capacity (Metric):
2.5 mL
8 mL
20 mL
50 mL
100 mL
200 mL
Description
Eliminates hazards of glass syringe breakage.
- Compatible with most syringe pumps
- Fully autoclavable for reuse
- Adaptable to Luer Lock or Swagelok™ fittings
- Resistant to most chemicals
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (Metric) | 8 mL |
| Diameter (English) Inside | 37/100 in. |
| Diameter (English) Outside | 1/2 in. |
| Diameter (Metric) Outside | 12.7 mm |
| Diameter (Metric) Inside | 9.3 mm |
| For Use With | Syringe Pump |
| IP Rating | None |
| Material | Stainless Steel (316) |
| Type | Swagelok 1/8 in. Syringe |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction