Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KDEL Antibody (1F2), Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | KDEL |
---|---|
Clone | 1F2 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000 |
Classification | Monoclonal |
Description
KDEL Monoclonal specifically detects KDEL in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
KDEL | |
1 mg/mL | |
Monoclonal | |
Purified | |
Cellular Markers, ER Markers, Membrane Trafficking and Chaperones | |
PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide | |
79070 | |
KDEL peptide (C-AVKDEL -COOH) conjugated at the N-terminal to Bovine Serum Albumin (BSA) | |
Primary | |
Detects KDEL tagged proteins. |
1F2 | |
Western Blot 1:1000 | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Endoplasmic reticulum resident protein 58, EP58KDEL1, ER protein 58, ERp58, KDEL (Lys-Asp-Glu-Leu) containing 1, KDEL motif-containing 1, KDEL motif-containing protein 1, MGC5302 | |
KDELC1 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title