Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KDEL Receptor Monoclonal Antibody (KR-10), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 11078100UG

This item is not returnable.
View return policy
Description
KDEL Receptor Monoclonal antibody specifically detects KDEL Receptor in Bovine, Canine, Chicken, Drosophila, Hamster, Human, Monkey, Mouse, Ovine, Porcine, Rabbit, Rat, Xenopus laevis samples. It is validated for Immunocytochemistry, Western BlotSpecifications
KDEL Receptor | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.2 | |
O18750, O76767, P06761, P08110, P08113, P11021, P14625, P20029, P24390, P33946, P34935, P41148, Q0VCX2, Q29092, Q569A6, Q66HD0, Q90593, Q91883, Q95M18, Q99JH8 | |
Mouse | |
Protein A | |
RUO | |
100008764, 100009456, 100349892, 100520962, 100689305, 100750912, 100773331, 101111703, 101122047, 10945, 14828, 22027, 25617, 282646, 3309, 34427, 361577, 362862, 374163, 396487, 397191, 397850, 404019, 407060, 415113, 480726, 484406, 618184, 68137, 7184, 780447 | |
2-8°C | |
Liquid |
Immunocytochemistry, Western Blot | |
KR-10 | |
Unconjugated | |
Hsp90b1 | |
78 kDa glucose-regulated protein; 8030486F04Rik; 94 kDa glucose-regulated protein; 94 kDa glucose-regulated protein {ECO:0000250; AL022860; AU019543; AW215843; baffled; Binding-immunoglobulin protein; BiP; D2Wsu141e; D2Wsu17e; ECGP; Endoplasmic reticulum chaperone BiP; endoplasmic reticulum lumenal Ca(2+)-binding protein grp78; endoplasmic reticulum resident protein 58; Endoplasmic reticulum resident protein 99; Endoplasmin; endoplasmin {ECO:0000250; endothelial cell (HBMEC) glycoprotein; EP58; epididymis luminal protein 35; epididymis secretory sperm binding protein Li 125m; epididymis secretory sperm binding protein Li 89n; ER lumen protein retaining receptor 1; ER lumen protein-retaining receptor 1; ER protein 58; ERD2; Erd2.1; ERp58; ERp99; glucose regulated protein, 78 kDa; glucose-regulated protein; glucose-regulated protein, 78kDa; gp96; gp96 homolog; GRP 78; Grp78; GRP-78; GRP94; GRP-94; HDEL; heat shock 70 kDa protein 5; heat shock 70kD protein 5; heat shock 70kD protein 5 (glucose-regulated protein, 78kD); heat shock 70kDa protein 5 (glucose-regulated protein); heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa); heat shock protein 5; Heat shock protein 70 family protein 5; heat shock protein 90 beta family member 1; heat shock protein 90 kDa beta member 1; heat shock protein 90 kDa beta member 1 {ECO:0000250; heat shock protein 90, beta (Grp94), member 1; heat shock protein 90, beta, member 1; heat shock protein 90kDa beta (Grp94), member 1; heat shock protein 90kDa beta family member 1; heat shock protein family A (Hsp70) member 5; Heat shock protein family A member 5; HEL35; HEL-S-125m; HEL-S-89n; Hsce70; HSP70 family protein 5; Hsp90b1; hsp90b1 {ECO:0000250; HSPA5; I79_019946; I79_020931; Immunoglobulin heavy chain-binding protein; KDEL; KDEL (Lys-Asp-Glu-Leu) containing 1; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1; KDEL endoplasmic reticulum protein retention receptor 1; KDEL motif containing 1; KDEL motif-containing 1; KDEL motif-containing protein 1; KDEL receptor 1; KDEL1; KDELC1; KDELR1; Lys-Asp-Glu-Leu; mBiP; MIF2; PM23; POGLUT2; Polymorphic tumor rejection antigen 1; Protein O-glucosyltransferase 2; Protein O-xylosyltransferase POGLUT2; Putative MAPK-activating protein PM23; RP23-446N16.1; Sez7; SEZ-7; Steroidogenesis-activator polypeptide; stress-inducible tumor rejection antigen gp96; TA-3; Targ2; Tra1; Tra-1; transforming growth factor alpha regulated gene 2; tumor rejection antigen (gp96) 1; tumor rejection antigen 1; Tumor rejection antigen gp96; UniProtKB:P08113}; UNQ1910/PRO4357; XAP-1 antigen | |
Peptide corresponding to aa 192-212 of bovine KDEL receptor | |
100 μg | |
Primary | |
Bovine, Canine, Chicken, Drosophila, Hamster, Human, Monkey, Mouse, Ovine, Pig, Rabbit, Rat, X. laevis | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction