Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KDELC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571393
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571393 Supplier Invitrogen™ Supplier No. PA571393
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KDELC1 Polyclonal Antibody for Western Blot

Protein glucosyltransferase that catalyzes the transfer of glucose from UDP-glucose to a serine residue within the consensus sequence peptide C-X-N-T-X-G-S-F-X-C. Can also catalyze the transfer of xylose from UDP-xylose but less efficiently. Specifically targets extracellular EGF repeats of proteins such as NOTCH1 and NOTCH3. May regulate the transport of NOTCH1 and NOTCH3 to the plasma membrane and thereby the Notch signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KDELC1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KDELC1
Gene Accession No. Q6UW63
Gene Alias endoplasmic reticulum resident protein 58; EP58; ER protein 58; ERp58; KDEL (Lys-Asp-Glu-Leu) containing 1; KDEL motif containing 1; KDEL motif-containing 1; KDEL motif-containing protein 1; KDEL1; KDELC1; POGLUT2; Protein O-glucosyltransferase 2; Protein O-xylosyltransferase POGLUT2; UNQ1910/PRO4357
Gene Symbols KDELC1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human KDELC1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79070
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.