Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KDELR1 Monoclonal Antibody (KR-10), PerCP
GREENER_CHOICE

Catalog No. PIMA545151
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545151 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545151 Supplier Invitrogen™ Supplier No. MA545151
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL was sufficient for detection of KDEL receptor in 20 μg monkey Vero cell lysate by colorimetric immunoblot analysis using Goat Anti-Mouse IgG:AP as the secondary. Detects approximately 25kDa.

KDELR1 (KDEL Endoplasmic Reticulum Protein Retention Receptor 1) is a Protein Coding gene. Diseases associated with KDELR1 include Syndromic X-Linked Intellectual Disability 94 and Myopathy, Centronuclear, 1. Among its related pathways are Transport to the Golgi and subsequent modification and Activation of cAMP-Dependent PKA. Gene Ontology (GO) annotations related to this gene include ER retention sequence binding and KDEL sequence binding. An important paralog of this gene is KDELR2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KDELR1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone KR-10
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.2
Gene KDELR1
Gene Accession No. P24390, P33946, Q569A6, Q99JH8
Gene Symbols KDELR1
Host Species Mouse
Immunogen A 21 residue synthetic peptide (amino acids 192-212) based on the bovine KDEL receptor and the peptide coupled to KLH.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100349892, 100520962, 100750912, 101122047, 10945, 361577, 484406, 618184, 68137
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Sheep, Hamster, Pig, Chicken, Chicken, Frog
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.