Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDELR1, Mouse anti-Bovine, ATTO488, Clone: KR-10, Abnova™

Catalog No. 89337972 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-972 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-972 Supplier Abnova Corporation Supplier No. MAB18047
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen KDELR1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone KR-10
Conjugate Atto 488
Dilution Immunocytochemistry (1:1000) Immunofluorescence (1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.09% sodium azide).
Gene KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1
Gene Alias MGC139648
Gene Symbols KDELR1
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 192-212 of bovine KDELR1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 618184
Target Species Bovine
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.