Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KDELR1, Mouse anti-Bovine, R-PE, Clone: KR-10, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB18057
Specifications
KDELR1 | |
Monoclonal | |
R-PE | |
In PBS, pH 7.2 (50% glycerol, 0.09% sodium azide). | |
MGC139648 | |
Mouse | |
100 μg | |
Primary | |
Bovine | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
KR-10 | |
Immunocytochemistry (1:1000) Immunofluorescence (1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1 | |
KDELR1 | |
A synthetic peptide (conjugated with KLH) corresponding to amino acids 192-212 of bovine KDELR1. | |
RUO | |
618184 | |
Store at -20°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction