Learn More
Invitrogen™ KDELR2 Polyclonal Antibody

Description
KDELR2 Polyclonal Antibody for ICC/IF, Western Blot
Specifications
Specifications
| Antigen | KDELR2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 |
| Gene | KDELR2 |
| Gene Accession No. | P33947, Q5U305, Q9CQM2 |
| Gene Alias | (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 2; 1110007A14Rik; ELP1; ELP-1; ER lumen protein retaining receptor 2; ER lumen protein-retaining receptor 2; ERD2.2; ERD22; ERD-2-like protein; ERD2-like protein 1; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 2; KDEL endoplasmic reticulum protein retention receptor 2; KDEL receptor 2; Kdelr2 |
| Gene Symbols | KDELR2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.