Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KDELR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560377
Description
Immunogen sequence: FYLYVTKGRS WDDSNADTGL RSYSSI Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 38%.
This gene encodes a member of the KDEL endoplasmic reticulum protein retention receptor family. Retention of resident soluble proteins in the lumen of the endoplasmic reticulum (ER) is achieved in both yeast and animal cells by their continual retrieval from the cis-Golgi, or a pre-Golgi compartment. Sorting of these proteins is dependent on a C-terminal tetrapeptide signal, usually lys-asp-glu-leu (KDEL) in animal cells, and his-asp-glu-leu (HDEL) in S. cerevisiae. This process is mediated by a receptor that recognizes, and binds the tetrapeptide-containing protein, and returns it to the ER. In yeast, the sorting receptor encoded by a single gene, ERD2, is a seven-transmembrane protein. Unlike yeast, several human homologs of the ERD2 gene, constituting the KDEL receptor gene family, have been described. KDELR3 was the third member of the family to be identified. Alternate splicing results in multiple transcript variants.
Specifications
KDELR3 | |
Polyclonal | |
Unconjugated | |
KDELR3 | |
AI173274; ER lumen protein retaining receptor 3; ER lumen protein-retaining receptor 3; ERD23; ERD2L3; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 3; KDEL endoplasmic reticulum protein retention receptor 3; KDEL receptor 3; Kdelr3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11015 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43731 | |
KDELR3 | |
Recombinant protein corresponding to Human KDELR3. Recombinant protein control fragment (Product #RP-100371). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction