Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KDF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566984
Description
Immunogen sequence: ANWAKEHNGVP PSPDRAPPSR RDGQRLKSTM GSSFSYPDVK LKGIPVYPYP RATSPAPDAD SCCKEPLADP PPMRHSL Highest antigen sequence identity to the following orthologs - mouse 88%, rat 90%.
Plays a role in the regulation of the epidermis formation during early development. Required both as an inhibitor of basal cell proliferation and a promoter of differentiation of basal progenitor cell progeny.
Specifications
KDF1 | |
Polyclonal | |
Unconjugated | |
KDF1 | |
1810019J16Rik; C1orf172; C2H1orf172; KDF1; Keratinocyte differentiation factor 1; RGD1303271; RP11-344H11.3; uncharacterized protein C1orf172 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126695 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NAX2 | |
KDF1 | |
Recombinant Human KDF1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction