Learn More
Invitrogen™ KDF1 Polyclonal Antibody

Description
Immunogen sequence: LDEQDAEGRL VRGIIRISTR KSRARPQTSE GRSTRAAAPT AAAPDSGHET MVGSGLSQDE LTVQISQETT ADAIARKLRP YGA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.
Specifications
Specifications
| Antigen | KDF1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | KDF1 |
| Gene Accession No. | Q8NAX2 |
| Gene Alias | 1810019J16Rik; C1orf172; C2H1orf172; KDF1; Keratinocyte differentiation factor 1; RGD1303271; RP11-344H11.3; uncharacterized protein C1orf172 homolog |
| Gene Symbols | KDF1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.