Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KDM1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 208131AP150UL

Description
LSD1 is a histone demethylase that specifically demethylates ′Lys-4′ of histone H3, a specific tag for epigenetic transcriptional activation, thereby acting as a corepressor. LSD1 contains a SWIRM domain, a FAD-binding motif, and an amine oxidase domain. This protein is a component of several histone deacetylase complexes, though it silences genes by functioning as a histone demethylase. It acts by oxidizing the substrate by FAD to generate the corresponding imine that is subsequently hydrolyzed. LSD1 demethylates both mono- and tri-methylted ′Lys-4′ of histone H3. This protein may play a role in the repression of neuronal genes. Alone, it is unable to demethylate H3 ′Lys-4′ on nucleosomes and requires the presence of RCOR1/CoREST to achieve such activity. It may also demethylate ′Lys-9′ of histone H3, a specific tag for epigenetic transcriptional repression, thereby leading to derepression of androgen receptor target genes. Mutations affecting this gene can result in cleft palate, psychomotor retardation, and distintive facial features.Specifications
KDM1 | |
Polyclonal | |
Unconjugated | |
KDM1A | |
AOF2, BHC110, KDM1, KDM1A, KIAA0601, LSD1, lysine (K) demethylase 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23028, 99982 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60341, Q6ZQ88 | |
Kdm1a | |
KDM1 Fusion Protein Ag14778 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction