Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDM1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980733
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0733 Supplier Bioss Supplier No. BS10166R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LSD1 is a histone demethylase that specifically demethylates ′Lys-4′ of histone H3, a specific tag for epigenetic transcriptional activation, thereby acting as a corepressor. LSD1 contains a SWIRM domain, a FAD-binding motif, and an amine oxidase domain. This protein is a component of several histone deacetylase complexes, though it silences genes by functioning as a histone demethylase. It acts by oxidizing the substrate by FAD to generate the corresponding imine that is subsequently hydrolyzed. LSD1 demethylates both mono- and tri-methylted ′Lys-4′ of histone H3. This protein may play a role in the repression of neuronal genes. Alone, it is unable to demethylate H3 ′Lys-4′ on nucleosomes and requires the presence of RCOR1/CoREST to achieve such activity. It may also demethylate ′Lys-9′ of histone H3, a specific tag for epigenetic transcriptional repression, thereby leading to derepression of androgen receptor target genes. Mutations affecting this gene can result in cleft palate, psychomotor retardation, and distintive facial features.

Specifications

Antigen KDM1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TBL1X
Gene Accession No. O60341, O60907
Gene Alias 1810043O07Rik; 5330429M20; AA408884; amine oxidase (flavin containing) domain 2; Aof2; BHC110; BOS_2431; BOS_26437; BRAF35-HDAC complex protein BHC110; CPRF; D4Ertd478e; EBI; FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit; F-box-like/WD repeat-containing protein TBL1X; Flavin-containing amine oxidase domain-containing protein 2; KDM1; KDM1A; KIAA0601; Lsd1; lysine (K)-specific demethylase 1; lysine (K)-specific demethylase 1A; lysine demethylase 1A; lysine-specific demethylase 1; lysine-specific histone demethylase 1; Lysine-specific histone demethylase 1A; mKIAA0601; neuroprotective protein 3; RGD1562975; RGD1563868; RP1-184J9.1; SMAP55; TBL1; Tbl1x; transducin (beta)-like 1 X-linked; transducin (beta)-like 1X-linked; transducin beta like 1 X-linked; transducin beta-like 1; Transducin beta-like protein 1X; Transducin-beta-like protein 1, X-linked; wu:fb82e06; wu:fc51a02; wu:fj36a05; wu:fj66b06; zgc:158242
Gene Symbols KDM1A, TBL1X
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KDM1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23028, 6907
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.