Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KDM1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572782
Description
KDM1B Polyclonal Antibody for Western Blot, ELISA
Flavin-dependent histone demethylases, such as KDM1B, regulate histone lysine methylation, an epigenetic mark that regulates gene expression and chromatin function.
Specifications
KDM1B | |
Polyclonal | |
Unconjugated | |
KDM1B | |
4632428N09Rik; AI482520; amine oxidase (flavin containing) domain 1; amine oxidase, flavin containing 1; Aof1; bA204B7.3; C6orf193; dJ298J15.2; Flavin-containing amine oxidase domain-containing protein 1; KDM1B; LSD2; lysine (K)-specific demethylase 1B; lysine demethylase 1B; lysine-specific histone demethylase 1B; Lysine-specific histone demethylase 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
218214, 221656 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8CIG3, Q8NB78 | |
KDM1B | |
KDM1B antibody for a 17 amino acid peptide near the amino terminus of human KDM1B. The immunogen is within amino acids 80 - 130 of KDM1B. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction