Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KDM2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144674
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144674 Supplier Invitrogen™ Supplier No. PA5144674
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: MCF-7 cell lysate, Jurkat cell lysate, PC-12 cell lysate, human brain tissue lysate, rat colon tissue, human stomach tissue, mouse colon tissue. Predicted band size: 133 kDa Subcellular Location: Nucleus, nucleoplasm, Chromosome.

FBXL11 is a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box). The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. FBXL11 belongs to the Fbls class and, in addition to an F-box, contains at least 6 highly degenerated leucine-rich repeats.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KDM2A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene KDM2A
Gene Accession No. P59997, Q9Y2K7
Gene Alias [Histone-H3]-lysine-36 demethylase 1A; 100043628; 5530401A10Rik; AA589516; AW536790; CXXC8; CXXC-type zinc finger protein 8; DKFZp434M1735; FBL11; FBL7; F-box and leucine-rich repeat protein 11; F-box protein containing leucine repeats; F-box protein FBL7; F-box protein Lilina; F-box/LRR-repeat protein 11; Fbxl11; FLJ00115; FLJ46431; Gm4560; Jhdm1; JHDM1A; JmjC domain-containing histone demethylation protein 1A; jumonji C domain-containing histone demethylase 1A; Kdm2a; KIAA1004; lalina; LILINA; lysine (K)-specific demethylase 2A; lysine demethylase 2A; lysine-specific demethylase 2A; SF-KDM2A
Gene Symbols KDM2A
Host Species Rabbit
Immunogen Recombinant protein within human KDM2A aa 381-580.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225876, 22992, 361700
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.