Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDM3A/JHDM2A (KDM3A) Monoclonal Antibody (OTI9F6), TrueMAB™, OriGene
SDP

Catalog No. 502543789
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3789 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3789 Supplier OriGene Supplier No. CF812140
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KDM3A/JHDM2A (KDM3A) Monoclonal antibody specifically detects KDM3A/JHDM2A (KDM3A) in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen KDM3A/JHDM2A (KDM3A)
Applications Western Blot
Classification Monoclonal
Clone OTI9F6
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene KDM3A
Gene Accession No. Q63679, Q6PCM1, Q9Y4C1
Gene Alias 1700105C21Rik; C230043E16Rik; DKFZp686A24246; DKFZp686P07111; JHDM2a; JHMD2A; JmjC domain-containing histone demethylation protein 2A; Jmjd1; Jmjd1a; jumonji C domain-containing histone demethylase 2A; jumonji domain containing 1A; jumonji domain-containing protein 1A; KDM2A; Kdm3a; Kiaa0742; lysine (K)-specific demethylase 3A; lysine demethylase 3A; lysine-specific demethylase 3A; probable zinc finger protein; testis specific gene A; testis-specific gene A protein; testis-specific protein A; TGSA; Tsga; zinc finger protein TSGA
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 850-1321 of human KDM3A (NP_060903) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104263, 312440, 55818
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.