Learn More
KDM3A/JMJD1A Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 128351AP150UL

Description
This antibody recognize the N-term of KDM3A (JMJD1A).
This gene encodes a zinc finger protein that contains a jumonji domain and may play a role in hormone-dependent transcriptional activation. Alternative splicing results in multiple transcript variants.Specifications
KDM3A/JMJD1A | |
Polyclonal | |
Unconjugated | |
KDM3A | |
JHDM2A, JHMD2A, JMJD1, JMJD1A, KDM3A, KIAA0742, lysine (K) demethylase 3A, Lysine specific demethylase 3A, TSGA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
104263, 55818 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6PCM1, Q9Y4C1 | |
KDM3A | |
KDM3A,JMJD1A Fusion Protein Ag3581 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.