Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDM3B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501732599 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2599 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2599 Supplier Proteintech Group Inc Supplier No. 199151AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone demethylase that specifically demethylates ′Lys-9′ of histone H3, thereby playing a central role in histone code. Demethylation of Lys residue generates formaldehyde and succinate. May have tumor suppressor activity.

Specifications

Antigen KDM3B
Applications Western Blot
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KDM3B
Gene Accession No. Q6ZPY7, Q7LBC6
Gene Alias 5qNCA, C5orf7, JHDM2B, JMJD1B, KDM3B, KIAA1082, lysine (K) demethylase 3B, Lysine specific demethylase 3B, NET22, Nuclear protein 5qNCA
Gene Symbols KDM3B
Host Species Rabbit
Immunogen KDM3B Fusion Protein Ag13795
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 277250, 51780
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.