Learn More
KDM4C Mouse anti-Human, Clone: OTI4B1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500587

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
JMJD2C is a member of the Jumonji domain 2 (JMJD2) family and encodes a protein with one JmjC domain, one JmjN domain, two PHD-type zinc fingers, and two Tudor domains. This nuclear protein functions as a trimethylation-specific demethylase, converting specific trimethylated histone residues to the dimethylated form. Chromosomal aberrations and increased transcriptional expression of this gene are associated with esophageal squamous cell carcinoma.Specifications
KDM4C | |
Monoclonal | |
Unconjugated | |
KDM4C | |
GASC1, JHDM3C, JMJD2C, TDRD14C | |
Mouse | |
Affinity Chromatography | |
RUO | |
23081 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI4B1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H3R0 | |
KDM4C | |
Full length human recombit protein of human KDM4C produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.