Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KDM4E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562939
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562939 Supplier Invitrogen™ Supplier No. PA562939
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GQGQGRGCSR GRGHGCCTRE LGTEEPTVQP ASKRRLLMGT RSRAQGHRPQ LPLANDLMTN LS Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 35%.

The protein encoded by this intronless gene is a member of a large family of histone lysine demethylases, which use oxygen and 2-oxoglutarate to demethylate di- and trimethylated lys9 of histone H3. Derepression of genes by demethylases is sometimes involved in viral infection or carcinogenesis, so inhibitors of these enzymes are desired.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KDM4E
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KDM4E
Gene Accession No. B2RXH2
Gene Alias JMJD2E; KDM4DL; Kdm4dl1; KDM4D-like protein; KDM4E; lysine (K)-specific demethylase 4D-like 1; lysine (K)-specific demethylase 4E; lysine demethylase 4E; lysine-specific demethylase 4D-like; Lysine-specific demethylase 4E
Gene Symbols KDM4E
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KDM4E. Recombinant protein control fragment (Product #RP-100858).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390245
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.