Learn More
Invitrogen™ KDM5A Recombinant Rabbit Monoclonal Antibody (5B10P1)

Description
Immunogen sequence: HAYSSASKSC SQGSSTPRKQ PRKSPLVPRS LEPPVLELSP GAKAQLEELM MVGDLLEVSL DETQHIWRIL QATHPPSEDR FLHIMEDDSM EEKPLKVKGK DSSEKKRKRK LEKVEQLFGE GKQKSKELKK MDKPRKKKLK LGADKSKELN KLAKKLAKEE ERKKKKEKAA AAKVELVKES TEKKREKKVL DIPSKYDWSG AEESDDENAV CAAQNCQRPC KDKVDWVQCD GGCDEWFHQV CVGVSPEMAE NEDYICINCA KKQGPVSPGP APPPSFIMSY KLPMEDLKET S.
Specifications
Specifications
| Antigen | KDM5A |
| Applications | ChIP Assay, ELISA, Immunoprecipitation, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 5B10P1 |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | KDM5A |
| Gene Accession No. | P29375, Q3UXZ9 |
| Gene Alias | AA409370; C76986; histone demethylase JARID1A; JARID1A; jumonji, AT rich interactive domain 1A (Rbp2 like); Jumonji, AT rich interactive domain 1A (RBP2-like); jumonji/ARID domain-containing protein 1A; Kdm5a; lysine (K)-specific demethylase 5A; lysine demethylase 5A; lysine-specific demethylase 5A; Rbbp2; RBBP-2; RBP2; retinoblastoma binding protein 2; retinoblastoma-binding protein 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.