Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KDM5B Monoclonal Antibody (CL1147)
GREENER_CHOICE

Catalog No. PIMA524627
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524627 Supplier Invitrogen™ Supplier No. MA524627
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: DGINSLERKL KRRLEREGLS SERWERVKKM RTPKKKKIKL SHPKDMNNFK LERERSYELV RSAETHSLPS DTSYSEQEDS E Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 91%. Binds to an epitope located within the peptide sequence SERWERVKKMRTPKK as determined by overlapping synthetic peptides.

KDM5B is a histone demethylase that demethylates 'Lys-4' of histone H3, thereby playing a central role in histone code. It does not demethylate histone H3 'Lys-9' or H3 'Lys-27'. KDM5B demethylates trimethylated, dimethylated and monomethylated H3 'Lys-4'. It acts as a transcriptional corepressor for FOXG1B and PAX9. It favors the proliferation of breast cancer cells by repressing tumor suppressor genes such as BRCA1 and HOXA5. In contrast, KDM5B may act as a tumor suppressor for melanoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KDM5B
Applications Western Blot
Classification Monoclonal
Clone CL1147
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KDM5B
Gene Accession No. Q9UGL1
Gene Alias 2010009J12Rik; 2210016I17Rik; AW556288; cancer/testis antigen 31; CT31; D1Ertd202e; Histone demethylase JARID1B; Jarid1b; jumonji, AT rich interactive domain 1B; jumonji, AT rich interactive domain 1B (Rbp2 like); Jumonji, AT rich interactive domain 1B (RBP2-like); jumonji/ARID domain-containing protein 1B; KDM5B; Kiaa4034; lysine (K)-specific demethylase 5B; lysine demethylase 5B; lysine-specific demethylase 5B; mKIAA4034; PLU1; PLU-1; PPP1R98; protein phosphatase 1, regulatory subunit 98; PUT1; putative DNA/chromatin binding motif; putative DNA/chromatin binding motif 1; Rb-Bp2; RBBP2H1; RBBP2H1A; RBP2-H1; Retinoblastoma-binding protein 2 homolog 1; retinoblastoma-binding protein 2, homolog 1A; RGD1565602
Gene Symbols KDM5B
Host Species Mouse
Immunogen Recombinant protein fragment of Human KDM5B (Product #RP-94108)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10765
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.