Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDM5B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987076
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7076 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7076 Supplier Bioss Supplier No. BS6139R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KDM5B is a histone demethylase that demethylates ′Lys-4′ of histone H3, thereby playing a central role in histone code. It does not demethylate histone H3 ′Lys-9′ or H3 ′Lys-27′. KDM5B demethylates trimethylated, dimethylated and monomethylated H3 ′Lys-4′. It acts as a transcriptional corepressor for FOXG1B and PAX9. It favors the proliferation of breast cancer cells by repressing tumor suppressor genes such as BRCA1 and HOXA5. In contrast, KDM5B may act as a tumor suppressor for melanoma.

Specifications

Antigen KDM5B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KDM5B
Gene Accession No. Q9UGL1
Gene Alias 2010009J12Rik; 2210016I17Rik; AW556288; Cancer/testis antigen 31; CT31; D1Ertd202e; Histone demethylase JARID1B; Jarid1b; jumonji, AT rich interactive domain 1B; jumonji, AT rich interactive domain 1B (Rbp2 like); Jumonji, AT rich interactive domain 1B (RBP2-like); Jumonji/ARID domain-containing protein 1B; Kdm5b; Kiaa4034; lysine (K)-specific demethylase 5B; lysine demethylase 5B; Lysine-specific demethylase 5B; mKIAA4034; Plu1; PLU-1; PPP1R98; protein phosphatase 1, regulatory subunit 98; PUT1; putative DNA/chromatin binding motif; putative DNA/chromatin binding motif 1; Rb-Bp2; RBBP2H1; RBBP2H1A; RBP2-H1; Retinoblastoma-binding protein 2 homolog 1; retinoblastoma-binding protein 2, homolog 1A; RGD1565602
Gene Symbols KDM5B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KDM5B/Jarid1B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10765
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.