Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KDM6A Monoclonal Antibody (6E2D6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548620
Description
KDM6A Monoclonal Antibody for Western Blot, ELISA
This gene is located on the X chromosome and is the corresponding locus to a Y-linked gene which encodes a tetratricopeptide repeat (TPR) protein. The encoded protein of this gene contains a JmjC-domain and catalyzes the demethylation of tri/dimethylated histone H3.
Specifications
KDM6A | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
O15550 | |
KDM6A | |
Purified recombinant fragment of human KDM6A (AA: 1252-1401) expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG2a |
ELISA, Western Blot | |
6E2D6 | |
Unconjugated | |
KDM6A | |
4lysine (K)-specific demethylase 6A; bA386N14.2; bA386N14.2 (ubiquitously transcribed X chromosome tetratricopeptide repeat protein (UTX)); histone demethylase UTX; KABUK2; Kdm6a; lysine (K)-specific demethylase 6A; lysine demethylase 6A; lysine-specific demethylase 6A; lysine-specific demethylase 6A-like protein; NEWGENE_1565481; ubiqiutous TPR motif protein; ubiquitous TPR motif protein UTX; ubiquitously transcribed tetratricopeptide repeat gene, X chromosome; ubiquitously transcribed tetratricopeptide repeat protein X-linked; ubiquitously transcribed tetratricopeptide repeat, X chromosome; ubiquitously transcribed TPR protein on the X chromosome; Ubiquitously transcribed X chromosome tetratricopeptide repeat protein; ubiquitously-transcribed TPR gene on the X chromosome; Ubiquitously-transcribed TPR protein on the X chromosome; Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein; UTX | |
Mouse | |
Protein G | |
RUO | |
103689969, 7403 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction