Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KDM6A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568598
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568598 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568598 Supplier Invitrogen™ Supplier No. PA568598
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Rabbit: 100%; Rat: 100% This antibody is 100% homologous to isoforms 1 and 2.

This gene is located on the X chromosome and is the corresponding locus to a Y-linked gene which encodes a tetratricopeptide repeat (TPR) protein. The encoded protein of this gene contains a JmjC-domain and catalyzes the demethylation of tri/dimethylated histone H3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KDM6A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KDM6A
Gene Accession No. O70546
Gene Alias 4lysine (K)-specific demethylase 6A; bA386N14.2; bA386N14.2 (ubiquitously transcribed X chromosome tetratricopeptide repeat protein (UTX)); histone demethylase UTX; KABUK2; Kdm6a; lysine (K)-specific demethylase 6A; lysine demethylase 6A; lysine-specific demethylase 6A; lysine-specific demethylase 6A-like protein; NEWGENE_1565481; ubiqiutous TPR motif protein; ubiquitous TPR motif protein UTX; ubiquitously transcribed tetratricopeptide repeat gene, X chromosome; ubiquitously transcribed tetratricopeptide repeat protein X-linked; ubiquitously transcribed tetratricopeptide repeat, X chromosome; ubiquitously transcribed TPR protein on the X chromosome; Ubiquitously transcribed X chromosome tetratricopeptide repeat protein; ubiquitously-transcribed TPR gene on the X chromosome; Ubiquitously-transcribed TPR protein on the X chromosome; Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein; UTX
Gene Symbols KDM6A
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human KDM6A (aa 245-294).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22289
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.