Learn More
Abnova™ Human KDR (NM_002253, 807 a.a. - 1356 a.a.) Partial Recombinant Protein with GST-tag expressed in Sf9 cells
Shop All Abnova Corporation ProductsDescription
Vascular endothelial growth factor (VEGF) is a major growth factor for endothelial cells. This gene encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. It functions as the main mediator of VEGF-induced endothelial proliferation, survival, migration, tubular morphogenesis and sprouting. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc.. Mutations of this gene are implicated in infantile capillary hemangiomas. [provided by RefSeq]
Specifications
Specifications
Accession Number | NM_002253 |
Concentration | 0.072 ug/uL |
For Use With (Application) | Functional Study, SDS-PAGE |
Formulation | Liquid |
Gene ID (Entrez) | 3791 |
Molecular Weight (g/mol) | 87.095kDa |
Name | KDR (Human) Recombinant Protein |
Preparation Method | Insect cell (Sf9) expression system |
Purification Method | GST-Affinity Chromatography |
Quality Control Testing | 2 ug/lane SDS-PAGE Stained with Coomassie Blue |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.